(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate

Supplier: Spectrum Chemical Mfg. Corp.

M1680-5GM M1680-25GM
700014-822EA 89.55 USD
700014-822 700014-820
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl-(S)-2-hydroxypropionate
Formula: C₁₃H₂₄O₃
Storage Temperature: Ambient
MDL Number: MFCD27977194
CAS Number: 61597-98-6

Order Now

Specification Test Results

Appearance White Crystallin
Melting Point 40°C min.
Assay (GC) 97% min.
Optical Rotation -79° to -83°


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR