IGEPAL® CA-720

Supplier: Spectrum Chemical Mfg. Corp.

I1149-500GM
700014-554EA 302.5 USD
700014-554
IGEPAL® CA-720
IGEPAL® CA-720

Igepal(R) CA-720 is a nonionic surfactant used in all phases of detergent compounding and aqueous processing in the textile and paper industries as well as industrial metal cleaners, acid cleaners, floor cleaners, detergent sanitizers and waterless hand cleaners.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₂₁(C₂H₄O)nOH
Density: 1.104
Storage Temperature: Ambient
MDL Number: MFCD00128254
CAS Number: 9036-19-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR