Hexane (mixture of isomers) ≥98.5% ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

H1010-100ML H1010-200LTB H1010-4LTGL H1010-20LTMT H1010-1LT H1010-500MLG
700014-468EA 83.5 USD
700014-468 700014-472 700014-476 700014-474 700014-470 700014-478
Hexane (mixture of isomers) ≥98.5% ACS
Hexane (mixture of isomers)

Hexanes, Reagent, ACS, is a mixture of hexane isomers plus methylcyclopentane and is a colorless liquid. It is a used as non-polar solvent since it is low cost, largely unreactive, and relatively safe nature and can be found in specialized glue, as a degreasing agent and to extract cooking oil from seeds. It is used in the laboratory to remove or extract oil and grease contaminant from soil and water sample for analysis. Because it cannot easily be deprotonated it is often used for reactions that involve very strong bases. As an ACS grade Reagent, Hexanes manufactured hexanes is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.672
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR