Glycerol monostearate, powder, Food Grade

Supplier: Spectrum Chemical Mfg. Corp.

GL149-12KG GL149-2.5KG GL149-45KGBL GL149-500GM
700014-460EA 1172.25 USD
700014-460 700014-462 700014-464 700014-466
Glycerol monostearate, powder, Food Grade
Glycerol monostearate

Glyceryl Monostearate, Powder, Food Grade is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color nor odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. In addition to use as an emulsifier, it is also an anti-caking and thickening agent.

Formula: C₂₁H₄₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00036186
CAS Number: 31566-31-1

Order Now

Specification Test Results

Monoglycerides Content 90.0 % min.
Free Glycerol (Diglycerol and Glycerol) max.1.2 %
Acid Value max. 3.0
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR