Glycerol monostearate, flakes

Supplier: Spectrum Chemical Mfg. Corp.

GL141-2.5KG GL141-45KGBL GL141-12KG GL141-500GM GL141-100GM
700014-454EA 390.7 USD
700014-454 700014-456 700014-452 700014-458 10860-434
Glycerol monostearate, flakes
Glycerol monostearate

Glyceryl Monostearate, Flake is commonly referred to as GMS and is an organic molecule that is often used as an emulsifier. It has neither color or odor but has a sweet taste and is a flaky hygroscopic powder. It is naturally occurring in the body and is the by product of fat breakdowns. It is used as a food additive and emulsifier, as well as an anti-caking and thickening agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₁H₄₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00036186
CAS Number: 31566-31-1

Order Now

Specification Test Results

Acid Value max. 3.0
Saponification Value 165 - 175
Iodine Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR