Ethyl benzoate ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Benzoic acid ethyl ester

E1034-500GM
700014-194EA 254.45 USD
700014-194
Ethyl benzoate ≥99%, Reagent Grade
Ethyl benzoate

Ethyl Benzoate, Reagent, an ester, is formed by the condensation of ethanol and benzoic acid and a component of several fragrances. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.051
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00009109
CAS Number: 93-89-0
EINECS: 202-284-3
Merck Index: 13,03802

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR