Carboxymethylcellulose calcium salt

Supplier: Spectrum Chemical Mfg. Corp.

C1528-25GM C1528-500GM C1528-100GM
700013-772EA 151.3 USD
700013-772 700013-774 700013-770
Carboxymethylcellulose calcium salt
Carboxymethylcellulose calcium salt

Carboxymethylcellulose Calcium Salt, also known as Carboxymethyl cellulose or cellulose gum, can be used as a thickening agent in pharmaceuticals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00197782
CAS Number: 9050-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR