Basic fuchsin

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline

BA135-25GM
700013-676EA 276.3 USD
700013-676
Basic fuchsin
Basic Fuchsin

Basic Fuchsin Hydrochloride, also known as rosaniline hydrochloride or fuchsin, is a dye that becomes magenta when dissolved in water.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
Melting Pt: 235 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012569
CAS Number: 632-99-5
EINECS: 211-189-6
Merck Index: 12,05686

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR