Gold metal, powder 20 Mesh for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

G1201-250MG G1201-5GM G1201-1GM
700013-272EA 311.75 USD
700013-272 700013-274 700013-270
Gold metal, powder 20 Mesh for synthesis
Gold

Gold Metal, 20 Mesh, Powder is a precious metal used as a surface coloring agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now

Specification Test Results

Particle size 20 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR