Dextran sulfate sodium salt, Powder

Supplier: Spectrum Chemical Mfg. Corp.

DE136-100GM DE136-25GM DE136-10KGBL
700013-168EA 1016.8 USD
700013-168 700013-174 700013-170
Dextran sulfate sodium salt, Powder
Dextran sulphate sodium salt

Dextran Sulfate Sodium Salt, Powder is a sodium salt of the sulfuric acid ester of Dextran with an average molecular weight of 40,000 - 50,000. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers. It appears as a white micro-crystalline powder that is soluble in cold water.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR