Dextran, M.W. ~ 40,000, powder suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP
DE124-25KGBL
DE124-25GM
DE124-1KG
DE124-100GM
700013-120EA
13254.8
USD
700013-120
700013-118
700013-116
700013-114
Dextran, M.W. ~ 40,000, powder suitable for use as excipient
Dextran
Dextran, 40,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 EINECS: 232-677-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...