Calcium alginate

Supplier: Spectrum Chemical Mfg. Corp.

C1674-100GM
700013-076EA 71.61 USD
700013-076 700013-074
Calcium alginate
Calcium alginate

Calcium Alginate can be used for forming artificial seeds in plant tissue culture and incorporated into wound dressings (alginate dressings) as a hemostatic.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00143567
CAS Number: 9005-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR