Methyl cellulose, 15 cP (2% H₂O) USP
Supplier: Spectrum Chemical Mfg. Corp.
M1314-2.5KG
M1314-500GM
M1314-12KG
M1314-45KGBL
M1314-100GM
95034-466EA
1270.7
USD
95034-466
95034-468
700013-028
700013-030
95034-464
Methyl cellulose, 15 cP (2% H₂O) USP
Methyl cellulose
Methylcellulose, USP is a bulk-forming laxative. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 |
Specification Test Results
Assay | 26% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...