Collodion, flexible USP

Supplier: Spectrum Chemical Mfg. Corp.
Danger

CO125-200LTB CO125-500MLG
700012-958EA 21707.95 USD
700012-958 95030-686 95030-694 95030-688
Collodion, flexible USP
Collodion

Flexible Collodion, USP is used as a surgical dressing and forms a flexible nitrocellulose membrane. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Boiling Pt: ∼34 °C (1013 hPa)
Melting Pt: 160 °C
Density: 0.77
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00081525
CAS Number: 9004-70-0
UN: 2059
ADR: 3,II
Merck Index: 13,02508

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR