Collodion USP

Supplier: Spectrum Chemical Mfg. Corp.
Danger

CO120-200LTB CO120-4LTGL CO120-100MLG CO120-500MLG
700012-954EA 16787.55 USD
700012-954 95030-696 700012-952 700012-956 95030-690 95030-692
Collodion USP
Collodion

Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Boiling Pt: ∼34 °C (1013 hPa)
Melting Pt: 160 °C
Density: 0.75 g/cm³ (20 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00081525
CAS Number: 9004-70-0
UN: 2059
ADR: 3,II
Merck Index: 13,02508

Order Now

Specification Test Results

Assay 5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR