Polyethyleneimine, M.W. ∼ 10,000 (by GPC), branched

Supplier: Spectrum Chemical Mfg. Corp.

P2996-25ML P2996-100ML P2996-250ML
700012-832EA 209.09 USD
700012-832 700012-826 700012-830
Polyethyleneimine, M.W. ∼ 10,000 (by GPC), branched
Polyethyleneimine

Polyethylenimine, Branched, Av. Mn (by GPC) Approx. 10,000, also known as Polyaziridine, has many applications in products like cosmetics, detergents, and adhesives as a chelating agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (CH2CH2NH)n
Density: 1.030
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR