Polyethyleneimine 50% (v/v) in aqueous solution

Supplier: Spectrum Chemical Mfg. Corp.

P1647-100ML
700012-802EA 236.41 USD
700012-802
Polyethyleneimine 50% (v/v) in aqueous solution
Polyethyleneimine

Polyethyleneimine, 50 Percent (v/v) Aqueous Solution, or PEI, is soluble in hot water as well as in methanol, ethanol, chloroform and at low pH. It ability to modify the surface of cellulose fibres make it usable in many products such as adhesives, cosmetics, water treatment agents and detergents

Formula: (CH2CH2NH)n
Density: 1.08
Storage Temperature: Ambient
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR