Acrylamide polymer

Supplier: Spectrum Chemical Mfg. Corp.

P2930-250GM P2930-50GM P2930-10GM P2930-500GM
700012-774EA 465.46 USD
700012-774 700012-778 700012-772 700012-776
Acrylamide polymer
Acrylamide polymer

Polyacrylamide, also called poly(1-carbamoylethylene), is used to flocculate solids in a liquid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: (CH₂:CHCONH₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084392
CAS Number: 9003-05-8

Order Now

Specification Test Results

Appearance White to Faintly
Loss On Drying max. 13%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR