Benzyl acetate ≥98%
Supplier: Spectrum Chemical Mfg. Corp.
Benzyl Acetate, an organic compound, is the ester formed by condensation of acetic acid and benzyl alcohol; it is the main constituent of the essential oils from jasmine flowers.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₁₀O₂ Boiling Pt: 206 °C (1013 hPa) Melting Pt: –51 °C Density: 1.04 Flash Pt: 95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008712 CAS Number: 140-11-4 EINECS: 205-399-7 Merck Index: 13,01125 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...