Acetaldehyde ≥99%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Danger

A1002-100ML
700011-592EA 731.71 USD
700011-592 700011-588 700011-590
Acetaldehyde ≥99%, Reagent Grade
Acetaldehyde

Acetaldehyde, Reagent is commercially used as a precursor to acetic acid. In the laboratory it is a reagent used for the detection of primary and secondary amines. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: CH₃CHO
Boiling Pt: 20.1 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.785
Flash Pt: –27 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006991
CAS Number: 75-07-0
EINECS: 200-836-8
UN: 1089
ADR: 3,I
Merck Index: 12,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de03379d171eda3c733a039cd16293e8ee4a02e2&vid=573943501bd6753f30bb12e41521b99361988998&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR