Starch (from wheat), powder

Supplier: Spectrum Chemical Mfg. Corp.

S1557-12KG
700011-354EA 871 USD
700011-354 700011-356 700011-358
Starch (from wheat), powder
Starch

Starch, Wheat, Powder is a carbohydrate and polysaccharide that has a large number of glucose units that are linked by glycosidic bonds derived from wheat. It is primarily used as a food additive and used in the fabrication of sweets such as wine gums and jelly beans where it is used as a mould. It is also used in the paper industry, clothing industry, as an adhesive and in the construction industry.

Formula: (C₆H₁₀O₅)n
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR