Peru balsam (Balsamum peruvianum)
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Peruvian Balsam is also known as Balsam of Peru and is derived from the tree grown in El Salvador which is known as Myroxylon. The balsam comes from the trunk of the tree which is similar to substances like vanilla, clove and cinnamon.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Density:
1.161 Storage Temperature: Ambient |
MDL Number:
MFCD00147893 CAS Number: 8007-00-9 |
Specification Test Results
Analysis By GC | To Pass Test |
Refractive Index @ 20°C | 1.5800 - 1.6000 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...