Lemongrass oil

Supplier: Spectrum Chemical Mfg. Corp.

L1092-500MLG L1092-100ML
700010-690EA 282.85 USD
700010-690 700010-688
Lemongrass oil
Lemongrass oil

Lemongrass Oil is considered an astringent and widely used in the perfume and soap industries.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00147429
CAS Number: 8007-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR