Eosin Y free acid

Supplier: Spectrum Chemical Mfg. Corp.

E1241-25GM
700010-284EA 338.3 USD
700010-284
Eosin Y free acid
Eosin Y

Eosin Y, Free Acid, also known as Solvent Red 43, is used as a personal care colorant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₈Br₄O₅
Melting Pt: 295 °C
Storage Temperature: Ambient
MDL Number: MFCD00036189
CAS Number: 15086-94-9
EINECS: 239-138-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR