Acid red 1

Supplier: Spectrum Chemical Mfg. Corp.

A3023-100GM
700009-986EA 182.55 USD
700009-986 700009-988
Acid red 1
Acid red 1

Azophloxine is a useful stain with color index of 18050.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₁₅N₃O₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00003954
CAS Number: 3734-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR