Berberine chloride ≥97.0%

Supplier: Spectrum Chemical Mfg. Corp.

B2777-25GM
700009-606EA 702.85 USD
700009-606 700009-610
Berberine chloride ≥97.0%
Berberine chloride

Berberine Chloride, also known as umbellatine, can reduce intracellular superoxide levels in LPS-stimulated macrophages.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₁₈ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD00011939
CAS Number: 633-65-8

Order Now

Specification Test Results

Assay 97.0 - 102.0 %
Water max. 16.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR