Theobromine

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: 3,7-Dimethyl-3,7-dihydro-1H-purine-2,6-dione, 3,7-Dimethylxanthine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3,7-dimethylpurine

TH105-25KGBL
700009-596EA 115.31 USD
700009-596 700009-598
Theobromine
Theobromine

Theobromine, formerly known as xantheose, is found in chocolate and classified as a xanthine alkaloid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₈N₄O₂
MW: 180.17 g/mol
Melting Pt: 350 °C
Density: 1.419 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00022830
CAS Number: 83-67-0
EINECS: 201-494-2
Merck Index: 12,09418

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR