Sinigrin hydrate

Supplier: Spectrum Chemical Mfg. Corp.

S1158-1GM
700009-548EA 400.51 USD
700009-548 700009-546
Sinigrin hydrate
Sinigrin hydrate

(-)-Sinigrin, Hydrate is used as a reference material in procedures for the isolation and identification of glucosinolates.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₇NO₉S₂
Storage Temperature: Ambient
MDL Number: MFCD00006616
CAS Number: 3952-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR