Asiaticoside ≥90%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (1S,2R,4aS,6aS,6bR,8aR,9R,10R,11R,12aR,12bR,14bS)-(2S,3R,4S,5S,6R)-6-((((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl 10,11-dihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate

A3649-100MG
700009-484EA 1230.55 USD
700009-484
Asiaticoside ≥90%
Asiaticoside

Asiaticoside is used to study potential treatments for wounds and burns. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄₈H₇₈O₁₉
Storage Temperature: Ambient
MDL Number: MFCD06642601
CAS Number: 16830-15-2

Order Now

Specification Test Results

Assay (HPLC) 90% min.
Loss On Drying max. 5%
Heavy Metals max. 20 ppm
Sulfated Ash max. 0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR