Morpholine ≥99% ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

M1340-20LT
700009-170EA 2391.95 USD
700009-170
Morpholine ≥99% ACS
Morpholine

Morpholine, Reagent, ACS is used as an industrial pH modifier and corrosion inhibitor. It is also used as organic synthetic building block and a laboratory solvent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₄H₉NO
Boiling Pt: 128.9 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.999
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00005972
CAS Number: 110-91-8
EINECS: 203-815-1
UN: 2054
ADR: 8.3,I
Merck Index: 12,06362
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9fb9ce2adfd770be2ae8d5f63ea6220bef0f781&vid=6b2bfa885631c3f4e0681813373d468dcc171b83&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR