Bromocresol green ACS

Supplier: Spectrum Chemical Mfg. Corp.

BR132-100GM BR132-25GM BR132-1KGBL BR132-5GM
700009-080EA 1242.75 USD
700009-080 700009-084 700009-082 700009-086
Bromocresol green ACS
Bromocresol green

Bromocresol Green, Reagent, ACS, is a dye in the triphenylmethane family and is used as a tracking dye and pH indicator for DNA agarose gel electrophoresis. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 13,01370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR