Polyvinylpyrrolidone, M.W. ∼ 10,000 K-15 for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

PO146-5KG PO146-500GM PO146-100GM
700008-882EA 1735.25 USD
700008-882 700008-880 700008-878
Polyvinylpyrrolidone, M.W. ∼ 10,000 K-15 for synthesis
Polyvinylpyrrolidone

Polyvinylpyrrolidone K-15 is also known as polyvidone or povidone. It has been used for trauma victims as a plasma volume expander, as well as a binder in some pharmaceutical tablets.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR