N-Methyl-2-pyrrolidone (NMP) ≥99% ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

M1280-200LTB
700008-852EA 7227.08 USD
700008-852 700008-854 700008-856 700008-858
N-Methyl-2-pyrrolidone (NMP) ≥99% ACS for synthesis
N-Methyl-2-pyrrolidone (NMP)

1-Methyl-2-pyrrolidone, Reagent, ACS is a laboratory analytical reagent that is miscible with water and several other solvents, including ethyl acetate, chloroform, benzene and alcohols and ketones. 1-METHYL-2-PYRROLIDINONE, REAGENT, ACS is colorless to slightly yellow, and is used in the processing of petrochemicals to recover pure hydrocarbons. It is also commonly used to dissolve a broad range of chemicals, in particular polymers. There are several industrial uses, including treating surfaces of textiles, resins and plastics, as well as for stripping paint. As an ACS grade Reagent, Spectrum Chemical manufactured 1-Methyl-2-pyrrolidinone is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.028
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR