Rhodamine B for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride

RH115-100GM RH115-25GM RH115-500GM
700008-556EA 116 USD
700008-556 700008-558 700008-560
Rhodamine B for synthesis
Rhodamine B

Rhodamine B, also known as Rhodamine 610, is used as a tracer dye within water to determine the direction and rate of transport and flow.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₈H₃₁ClN₂O₃
Melting Pt: 199…201 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011931
CAS Number: 81-88-9
EINECS: 201-383-9
Merck Index: 13,08266

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR