Fluorescein, alcohol soluble

Supplier: Spectrum Chemical Mfg. Corp.

FL113-100GM
700008-490EA 302.5 USD
700008-490 700008-492
Fluorescein, alcohol soluble
Fluorescein

Fluorescein, Alcohol Soluble is a fluorophore commonly used in microscopy, in a type of dye laser as the gain medium, in forensics and serology to detect latent blood stains, and in dye tracing.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₁₂O₅
Melting Pt: 314…316 °C
Storage Temperature: Ambient
MDL Number: MFCD00005050
CAS Number: 2321-07-5
EINECS: 219-031-8
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97b9d8d37f0106f4c8ef2d7759759d21bd43cc03&vid=25b78df95a04ab26c5ba8c96b95f031056926b13&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b0e97bec9c9f44dd58e1cd0ca79d7e94c3ba82d&vid=a29b350acb5fd71b32a30baa408af54494e42d59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR