(-)-Epigallocatechin gallate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate, (2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate, EGCG

E1154-50MG
700008-480EA 115.35 USD
700008-480
(-)-Epigallocatechin gallate
(-)-Epigallocatechin gallate

(-)-Epigallocatechin Gallate is an antioxidant polyphenol flavonoid isolated from green tea. Its possible benefit as a nutritional chemopreventive agent for cancer, atherosclerosis, and neurodegenerative diseases is generating increased scientific interest. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00075940
CAS Number: 989-51-5

Order Now

Specification Test Results

Appearance Cream Colored Po
Purity 95% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR