Ellagic acid ≥97%

Supplier: Spectrum Chemical Mfg. Corp.

E2173-5GM
700008-364EA 280.85 USD
700008-364
Ellagic acid ≥97%
Ellagic acid

Ellagic Acid, also known as 4,4′,5,5′,6,6′-Hexahydroxydiphenic acid, is a phenol antioxidant found in several vegetables and fruits. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.


GHS hazard : Exclamation mark

Formula: C₁₄H₆O₈
Storage Temperature: Ambient
MDL Number: MFCD00006914
CAS Number: 476-66-4

Order Now

Specification Test Results

Assay Approx. 97 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR