L(-)-Glutathione (oxidised form) ≥98%, oxidized
Supplier: Spectrum Chemical Mfg. Corp.
L-Glutathione, Oxidized, also referred to as GSH and is an organic chemical that can be traced in plants and animals alike. It is an antioxidant that prevents damage from reactive oxygen species to cellular components that are important. It can be used to raise levels of plasma and liver GSH concentrations and is thought to have some positive effects on reducing cancer development.
Formula:
C₂₀H₃₂N₆O₁₂S₂ Melting Pt: 185 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063106 CAS Number: 27025-41-8 EINECS: 248-170-7 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...