D-Saccharic acid potassium salt ≥98.5%

Supplier: Spectrum Chemical Mfg. Corp.

S1468-100GM
700007-866EA 1127.3 USD
700007-866 700007-868
D-Saccharic acid potassium salt ≥98.5%
D-Saccharic acid potassium salt

D-Saccharic Acid Monopotassium Salt, also known as glucaric acid, is derived by oxidizing a sugar with nitric acid. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: C₆H₉KO₈
Storage Temperature: Ambient
CAS Number: 576-42-1

Order Now

Specification Test Results

Assay 98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR