Tetradecyltrimethylammonium bromide ≥99%, Reagent Grade for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
Synonyms:
MiTMAB, (1-Tetradecyl)trimethylammonium bromide, N,N,N-Trimethyltetradecan-1-aminium bromide, Tetradonium bromide, Tetradecyl trimethyl ammonium Bromide, Myristyltrimethylammonium bromide, Dynamin Inhibitor II, TTAB
T1027-50KGBL
T1027-12KGBL
T1027-500GM
700007-646EA
3073.05
USD
700007-646
700007-640
700007-644
Tetradecyltrimethylammonium bromide ≥99%, Reagent Grade for synthesis
Tetradecyltrimethylammonium bromide
Tetradecyltrimethylammonium Bromide, Reagent
Formula:
C₁₇H₃₈BrN MW: 336.394 g/mol Melting Pt: 245…250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011770 CAS Number: 1119-97-7 EINECS: 214-291-9 UN: 1759 ADR: 8 II Merck Index: 12,06419 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...