Benzyldimethylhexadecylammonium chloride ≥97%

Supplier: Spectrum Chemical Mfg. Corp.

B2875-100GM
700007-558EA 168.9 USD
700007-558
Benzyldimethylhexadecylammonium chloride ≥97%
Benzyldimethylhexadecylammonium chloride
Formula: C₂₅H₄₆ClN
Storage Temperature: Ambient
MDL Number: MFCD00011625
CAS Number: 122-18-9
EINECS: 204-526-3

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR