Betaine, anhydrous

Supplier: Spectrum Chemical Mfg. Corp.

B1572-2.5KG B1572-25KGBL B1572-100GM B1572-500GM
700007-546EA 663.85 USD
700007-546 700007-548 700007-544 700007-550
Betaine, anhydrous
Betaine

Betaine, Anhydrous is an organic compound found mostly in plants and is considered an N-trimethylated amino acid. The addition of betaine to polymerase chain (PCR) reactions improves the amplification of DNA by reducing the formation of secondary structure in GC-rich regions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD00012123
CAS Number: 107-43-7
EINECS: 203-490-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR