EDTA (ethylenediamine tetraacetic acid) ≥98% NF

Supplier: Spectrum Chemical Mfg. Corp.
Warning

E1208-25KGBL
700007-312EA 2998.35 USD
700007-312 95033-076 95033-074
EDTA (ethylenediamine tetraacetic acid) ≥98% NF
EDTA (Ethylenediamine tetraacetic acid)

Edetic Acid, NF is used as a chelating agent in therapy and as preservative adjuncts. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₀H₁₆N₂O₈
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR