EDTA tetrasodium salt dihydrate ≥98%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Danger
|
E1047-12KG
E1047-3KG
E1047-500GM
E1047-125GM
700007-296EA
1278.59
USD
700007-296
700007-298
700007-302
700007-294
EDTA tetrasodium salt dihydrate ≥98%, Reagent Grade
EDTA tetrasodium salt dihydrate
Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent is a solid that has no color and is water soluble. It is mainly used to dissolve lime scale but has many other uses in fields such as medicine, industry, cosmetics and laboratories as a chelating agent and sequestrant. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₁₀H₁₂N₂Na₄O₈·2H₂O Melting Pt: > 300 °C Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150026 CAS Number: 10378-23-1 EINECS: 200-573-9 Merck Index: 13,03544 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...