2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid ≥90%

Supplier: Spectrum Chemical Mfg. Corp.

T1374-250ML
700006-642EA 187.14 USD
700006-642 700006-638 700006-640
2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid ≥90%
2,2'-[Oxybis(2,1-ethanediyloxy)]bisacetic acid

3,6,9-Trioxaundecanedioic Acid is used as an exfoliating, anti-aging, moisturizing, conditioning and emulsifying agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₈H₁₄O₇
Density: 1.3
Storage Temperature: Ambient
MDL Number: MFCD00044099
CAS Number: 13887-98-4

Order Now

Specification Test Results

Assay Approx. 90 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR