Potassium hydrogen phthalate ≥99.95%, crystals ACS, Reagent Special for acid-base titration

Supplier: Spectrum Chemical Mfg. Corp.

P1190-125GM P1190-2.5KG P1190-25GM P1190-500GM P1190-12KG P1190-50KGBL
700006-006EA 205.94 USD
700006-006 700006-010 700006-012 700006-014 700006-008 700006-016 700006-018
Potassium hydrogen phthalate ≥99.95%, crystals ACS, Reagent Special for acid-base titration
Potassium hydrogen phthalate

Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS is used in analytical chemistry as a disintegrating agent can be used as a buffering agent (in combination with hydrochloric acid (HCl) or sodium hydroxide (NaOH). As an ACS grade Reagent, Spectrum Chemical manufactured Potassium Hydrogen Sulfate is used as the quality standard against which other Potassium Hydrogen Sulfate substances are graded. it has met the toughest regulatory standards for quality and pureness

Formula: HOOCC₆H₄COOK
Melting Pt: 295…300 °C
Density: 1.6362 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013070
CAS Number: 877-24-7
EINECS: 212-889-4
Merck Index: 13,07694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4652479e822d1591ac7eb5e29f892b8c94588ad4&vid=75071da7f30b07f7bd077699a3b54ec89d80f0b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR