Potassium hydrogen phthalate ≥99.95%, crystals ACS, Reagent Special for acid-base titration
Supplier: Spectrum Chemical Mfg. Corp.
Potassium Hydrogen Phthalate, Acidimetric Standard, Crystal, Reagent Special, ACS is used in analytical chemistry as a disintegrating agent can be used as a buffering agent (in combination with hydrochloric acid (HCl) or sodium hydroxide (NaOH). As an ACS grade Reagent, Spectrum Chemical manufactured Potassium Hydrogen Sulfate is used as the quality standard against which other Potassium Hydrogen Sulfate substances are graded. it has met the toughest regulatory standards for quality and pureness
Formula:
HOOCC₆H₄COOK Melting Pt: 295…300 °C Density: 1.6362 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013070 CAS Number: 877-24-7 EINECS: 212-889-4 Merck Index: 13,07694 |
|
Specification Test Results
Assay | 99.95% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...