Diethyl phthalate ≥98% NF

Supplier: Spectrum Chemical Mfg. Corp.

D1097-200LTB D1097-20LT D1097-4LTGL D1097-500MLG D1097-500MLG
700006-000EA 12028.05 USD
700006-000 700006-002 95032-898 95032-900 700006-004
Diethyl phthalate ≥98% NF
Diethyl phthalate

Diethyl Phthalate, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₁₂H₁₄O₄
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: –3 °C
Density: 1.121
Flash Pt: 156 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d0a6c6b14ead75148c0661141b5e8d00e6fc1a9&vid=2461e9f0e230ca44b5372d1233410fbf47a564c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR