Disodium fumarate

Supplier: Spectrum Chemical Mfg. Corp.

F2075-100GM F2075-500GM
700005-916EA 180.45 USD
700005-916 700005-918 700005-920
Disodium fumarate
Disodium fumarate

Fumaric Acid Disodium Salt, similar to fumaric acid, is a white crystalline salt and has been evaluated as a food additive.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₄H₂Na₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00064567
CAS Number: 17013-01-3
EINECS: 241-087-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR