Phenolphthalein, powder ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

P1075-125GM P1075-50KGBL P1075-25GM P1075-2.5KG P1075-500GM
700005-602EA 263.73 USD
700005-602 700005-610 700005-606 700005-604 700005-608
Phenolphthalein, powder ACS for synthesis
Phenolphthalein

Phenolphthalein, Powder, Reagent, ACS is a chemical compound that varies in color according to the acidity level of the solution it is in. It has traditionally been used as a laxative but has more recently been used as part of a test to identify blood in substances, known as the Kastle-Mayer test. It is also used in certain toys to create disappearing ink effect. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: C₂₀H₁₄O₄
Melting Pt: 258…262 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
REACH: 01-2119498295-24
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR